Gene Transcripts

Gene Symbol
APLF
Entrez Gene ID
200558
Gene Name
aprataxin and PNKP like factor
Gene Aliases
APFL, C2orf13, PALF, Xip1, ZCCHH1
Chromosome Location
Chr. 2: 68467559 - 68580162 on Build GRCh38
UniGene ID
Hs.720369
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173545.2 NP_775816.1 6 961
GenBank mRNA AK290841.1 - 6 961
AK308186.1 - 5 851
BC041144.1 - 6 910 6042653

View Full Product Details