Gene Transcripts

Gene Symbol
C15orf52
Entrez Gene ID
388115
Gene Name
chromosome 15 open reading frame 52
Gene Aliases
Chromosome Location
Chr. 15: 40331452 - 40340967 on Build GRCh38
UniGene ID
Hs.32433
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207380.2 NP_997263.2 11 4320
GenBank mRNA AK124643.1 - 1 2333
AK125329.1 - 13 3070
AK126035.1 - 7 3703
BM716672.1 - 1 453
BQ690903.1 - 1 195
CR749408.1 - 8 4013

View Full Product Details