Gene Transcripts

Gene Symbol
TEX43
Entrez Gene ID
389320
Gene Name
testis expressed 43
Gene Aliases
C5orf48, Tseg7
Chromosome Location
Chr. 5: 126631722 - 126636282 on Build GRCh38
UniGene ID
Hs.177983
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207408.1 NP_997291.1 2 144
GenBank mRNA AK131015.1 - 2 144
BC130495.1 - 2 152 40146526
BC130497.1 - 2 152 40146528

View Full Product Details