Gene Transcripts

Gene Symbol
RAB44
Entrez Gene ID
401258
Gene Name
RAB44, member RAS oncogene family
Gene Aliases
RASD3, RASL13, dJ431A14.3
Chromosome Location
Chr. 6: 36697808 - 36733183 on Build GRCh38
UniGene ID
Hs.485258
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001257357.1 NP_001244286.1 14 3429
GenBank mRNA AK125083.1 - 6 983

View Full Product Details