Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008964.4 | NP_032990.1 | 2 | 2299 | |
GenBank mRNA | AK036531.1 | - | 2 | 2217 | |
AK054132.1 | - | 2 | 1820 | ||
AK086904.1 | - | 2 | 2214 | ||
AK136577.1 | - | 2 | 1312 | ||
AK137515.1 | - | 2 | 1509 | ||
BC005440.1 | - | 2 | 1807 | 3487087 | |
D50589.1 | - | 2 | 1587 |