Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009184.2 | NP_033210.1 | 4, 5 | 950 | |
GenBank mRNA | AK154717.1 | - | 688 | ||
BC125342.1 | - | 4, 5 | 921 | 40129857 | |
BC125344.1 | - | 4, 5 | 921 | 40129859 | |
BC145042.1 | - | 4, 5 | 890 | ||
U16805.1 | - | 4, 5 | 950 |