Gene Transcripts

Gene Symbol
Sstr4
Entrez Gene ID
20608
Gene Name
somatostatin receptor 4
Gene Aliases
Smstr4, sst4
Chromosome Location
Chr. 2: 148395377 - 148396764 on Build GRCm38
UniGene ID
Mm.35324
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009219.3 NP_033245.2 1 890
GenBank mRNA AK051189.1 - 925
BC138487.1 - 869 8861507
BC138488.1 - 869 8861508

View Full Product Details