Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253883.1 | NP_001240812.1 | 11 | 2061 | |
NM_011099.3 | NP_035229.2 | 11 | 2061 | ||
GenBank mRNA | AK002341.1 | - | 1 | 265 | |
AK135397.1 | - | 11 | 1854 | ||
AK151724.1 | - | 11 | 1927 | ||
AK153483.1 | - | 11 | 1927 | ||
AK154695.1 | - | 11 | 1853 | ||
AK155110.1 | - | 11 | 1854 | ||
AK168943.1 | - | 11 | 1854 | ||
AK170892.1 | - | 11 | 1857 | ||
AK171023.1 | - | 11 | 1854 | ||
AK171033.1 | - | 11 | 1857 | ||
AK178610.1 | - | 1 | 229 | ||
AK203758.1 | - | 1 | 229 | ||
AK204827.1 | - | 1 | 228 | ||
BB819292.1 | - | 1 | 44 | ||
BC016619.1 | - | 11 | 1821 | 3598842 | |
BC094663.1 | - | 11 | 1800 | 6829532 | |
D38379.1 | - | 11 | 1795 | ||
X97047.1 | - | 10 | 1772 |