Gene Transcripts

Gene Symbol
Sdc3
Entrez Gene ID
20970
Gene Name
syndecan 3
Gene Aliases
Synd3, mKIAA0468, syn-3
Chromosome Location
Chr. 4: 130792537 - 130826318 on Build GRCm38
UniGene ID
Mm.206536
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011520.3 NP_035650.2 4 1225
GenBank mRNA AK045898.1 - 4 1011
AK129153.1 - 4 1225
AK150120.1 - 2 324
AK155127.1 - 4 1210
BC054795.1 - 4 1184 6834272
BC062093.1 - 4 1121 5705893

View Full Product Details