Gene Transcripts

Gene Symbol
Nxnl1
Entrez Gene ID
234404
Gene Name
nucleoredoxin-like 1
Gene Aliases
A930031O08Rik, RdCVF, Txnl6
Chromosome Location
Chr. 8: 71560555 - 71568666 on Build GRCm38
UniGene ID
Mm.213208
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145598.2 NP_663573.1 1 158
GenBank mRNA BC017153.1 - 1 132
BC021911.1 - 1 110 4502967
BI738445.1 - 1 158

View Full Product Details