Gene Transcripts

Gene Symbol
St8sia6
Entrez Gene ID
241230
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 6
Gene Aliases
1700007J08Rik, AI314453, AI875066, ST8SiaVI, Siat8f
Chromosome Location
Chr. 2: 13651018 - 13794734 on Build GRCm38
UniGene ID
Mm.330004
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145838.1 NP_665837.1 5 542
GenBank mRNA AB059554.1 - 5 542
AK085105.1 - 6 679
AK156557.1 - 5 1079
AK172400.1 - 5 607
BC115912.1 - 5 497 40104779
BC118063.1 - 5 497 40104780

View Full Product Details