Gene Transcripts

Gene Symbol
Btbd7
Entrez Gene ID
238386
Gene Name
BTB (POZ) domain containing 7
Gene Aliases
5730507E09Rik, 8030448M07, E130118E17Rik, FUP1, mKIAA1525
Chromosome Location
Chr. 12: 102780797 - 102878471 on Build GRCm38
UniGene ID
Mm.397654
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172806.2 NP_766394.2 4 887
GenBank mRNA AK033160.1 - 3 800
AK122522.1 - 2 537
AK134271.1 - 3 738
BC041669.1 - 4 887 3482657
BC057303.1 - 3 606 5702276

View Full Product Details