Gene Transcripts

Gene Symbol
Lrrn1
Entrez Gene ID
16979
Gene Name
leucine rich repeat protein 1, neuronal
Gene Aliases
2810047E21Rik, NLRR-1
Chromosome Location
Chr. 6: 107529726 - 107570228 on Build GRCm38
UniGene ID
Mm.428543
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008516.4 NP_032542.1 2 935
GenBank mRNA AK139355.1 - 2 893
AK139965.1 - 2 937
AK160841.1 - 2 814
AK164849.1 - 2 894
AK173186.1 - 1 180
BC031122.1 - 1 210 4972803
BC058701.1 - 2 868 6853964
D45913.1 - 2 881

View Full Product Details