Gene Transcripts

Gene Symbol
Alk
Entrez Gene ID
11682
Gene Name
anaplastic lymphoma kinase
Gene Aliases
CD246, Tcrz
Chromosome Location
Chr. 17: 71868988 - 72605410 on Build GRCm38
UniGene ID
Mm.311854
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007439.2 NP_031465.2 3 1548
GenBank mRNA AK051358.1 - 3 1549
AK158763.1 - 3 1870

View Full Product Details