Gene Transcripts

Gene Symbol
Ap1s1
Entrez Gene ID
11769
Gene Name
adaptor protein complex AP-1, sigma 1
Gene Aliases
AP19
Chromosome Location
Chr. 5: 137034993 - 137046060 on Build GRCm38
UniGene ID
Mm.833
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007457.2 NP_031483.1 3 304
GenBank mRNA AK002225.1 - 3 282
BC052692.1 - 3 308 5714625
M62418.1 - 3 304

View Full Product Details