Gene Transcripts

Gene Symbol
Bub1
Entrez Gene ID
12235
Gene Name
budding uninhibited by benzimidazoles 1 homolog (S. cerevisiae)
Gene Aliases
AL022991, Bub1a, C80208, D2Xrf87
Chromosome Location
Chr. 2: 127800200 - 127831876 on Build GRCm38
UniGene ID
Mm.2185
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113179.1 NP_001106650.1 2, 3 208
NM_009772.2 NP_033902.2 2, 3 208
GenBank mRNA AF002823.1 - 2, 3 154
AK021258.1 - 2, 3 199
AK042932.1 - 2, 3 209
AK082540.1 - 2, 3 154
AK144213.1 - 2, 3 217
BC029116.1 - 2, 3 174 3671932
U89795.1 - 2, 3 226

View Full Product Details