Gene Transcripts

Gene Symbol
Bub1
Entrez Gene ID
12235
Gene Name
budding uninhibited by benzimidazoles 1 homolog (S. cerevisiae)
Gene Aliases
AL022991, Bub1a, C80208, D2Xrf87
Chromosome Location
Chr. 2: 127800200 - 127831876 on Build GRCm38
UniGene ID
Mm.2185
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113179.1 NP_001106650.1 5 555
NM_009772.2 NP_033902.2 5 555
GenBank mRNA AF002823.1 - 5 501
AK021258.1 - 5 546
AK042932.1 - 5 556
AK082540.1 - 5 501
AK144213.1 - 5 564
BC029116.1 - 5 521 3671932
U89795.1 - 5 573

View Full Product Details