Gene Transcripts
- Gene Symbol
- Capn2
- Entrez Gene ID
- 12334
- Gene Name
- calpain 2
- Gene Aliases
- AI326419, Capa-2, Capa2, m-calpain, m-calpin
- Chromosome Location
- Chr. 1: 182467259 - 182517483 on Build GRCm38
- UniGene ID
- Mm.19306
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009794.3 |
NP_033924.2
|
6, 7 |
947 |
|
GenBank mRNA |
AF015038.1 |
- |
6, 7 |
893 |
|
AK052141.1 |
- |
6, 7 |
922 |
|
AK053832.1 |
- |
6, 7 |
2233 |
|
AK078787.1 |
- |
2, 3 |
1576 |
|
AK150695.1 |
- |
6, 7 |
921 |
|
AK152094.1 |
- |
6, 7 |
921 |
|
AK152790.1 |
- |
6, 7 |
923 |
|
AK152994.1 |
- |
6, 7 |
923 |
|
AK154855.1 |
- |
6, 7 |
961 |
|
BC054726.1 |
- |
6, 7 |
947 |
5703563 |
D38117.1 |
- |
6, 7 |
988 |
|
Y10139.1 |
- |
6, 7 |
829 |
|
View Full Product Details