Gene Transcripts

Gene Symbol
Rnpepl1
Entrez Gene ID
108657
Gene Name
arginyl aminopeptidase (aminopeptidase B)-like 1
Gene Aliases
1110014H17Rik, AI894167
Chromosome Location
Chr. 1: 92910825 - 92920585 on Build GRCm38
UniGene ID
Mm.200971
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181405.4 NP_852070.3 11 2468
GenBank mRNA AK075626.1 - 7 1734
AK087430.1 - 5 972
AK138067.1 - 401
AK154881.1 - 8 3788
AK186285.1 - 1 58
AK204769.1 - 1 97
AV008848.2 - 1 115
BC024079.1 - 5 981
BC031789.1 - 11 1888 4006916

View Full Product Details