Gene Transcripts
- Gene Symbol
- Olfr6
- Entrez Gene ID
- 233670
- Gene Name
- olfactory receptor 6
- Gene Aliases
- M50, MOR103-16
- Chromosome Location
- Chr. 7: 106955984 - 106956934 on Build GRCm38
- UniGene ID
- Mm.296765
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC127998.1 |
- |
1 |
483 |
|
View Full Product Details