Gene Transcripts
- Gene Symbol
- RBM10
- Entrez Gene ID
- 8241
- Gene Name
- RNA binding motif protein 10
- Gene Aliases
- DXS8237E, GPATC9, GPATCH9, S1-1, TARPS, ZRANB5
- Chromosome Location
- Chr. X: 47145196 - 47186815 on Build GRCh38
- UniGene ID
- Hs.401509
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001204466.1 |
NP_001191395.1
|
10, 11 |
1319 |
|
NM_001204467.1 |
NP_001191396.1
|
11, 12 |
1547 |
|
NM_001204468.1 |
NP_001191397.1
|
11, 12 |
1581 |
|
NM_005676.4 |
NP_005667.2
|
11, 12 |
1550 |
|
NM_152856.2 |
NP_690595.1
|
10, 11 |
1316 |
|
XM_005272677.4 |
XP_005272734.1
|
11, 12 |
1600 |
|
XM_005272678.4 |
XP_005272735.1
|
10, 11 |
1134 |
|
XM_005272679.4 |
XP_005272736.1
|
10, 11 |
1131 |
|
XM_006724563.1 |
XP_006724626.1
|
7, 8 |
767 |
|
XM_011543989.2 |
XP_011542291.1
|
3, 4 |
491 |
|
XM_017029884.1 |
XP_016885373.1
|
10, 11 |
1470 |
|
XM_017029885.1 |
XP_016885374.1
|
10, 11 |
1242 |
|
GenBank mRNA |
AK292758.1 |
- |
10, 11 |
1270 |
|
AL137421.1 |
- |
12, 13 |
1439 |
|
BC000681.2 |
- |
8, 9 |
772 |
|
BC003089.1 |
- |
10, 11 |
1255 |
3536058 |
BC004181.2 |
- |
11, 12 |
1503 |
2961094 |
BC008733.2 |
- |
11, 12 |
1535 |
3163064 |
BC024153.2 |
- |
10, 11 |
1165 |
3845258 |
BX537969.1 |
- |
11, 12 |
1532 |
|
D50912.1 |
- |
11, 12 |
1398 |
|
KU178411.1 |
- |
9, 10 |
923 |
|
KU178412.1 |
- |
10, 11 |
1157 |
|
View Full Product Details