Gene Transcripts

Gene Symbol
Trav16d-dv11
Entrez Gene ID
547329
Gene Name
T cell receptor alpha variable 16D-DV11
Gene Aliases
ENSMUSG00000072535, ENSMUSG00000076796, Gm10893, Trav16d/dv11
Chromosome Location
Chr. 14: 53047287 - 53047821 on Build GRCm38
UniGene ID
Mm.321046
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK088475.1 - 751

Gene Symbol
C920008G01Rik
Entrez Gene ID
320734
Gene Name
RIKEN cDNA C920008G01 gene
Gene Aliases
TRAV7D-6
Chromosome Location
Chr. 14: 53020459 - 53020991 on Build GRCm38
UniGene ID
Mm.321046
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK050592.1 - 416

View Full Product Details