Gene Transcripts
- Gene Symbol
- Spice1
- Entrez Gene ID
- 212514
- Gene Name
- spindle and centriole associated protein 1
- Gene Aliases
- Ccdc52, D16Ertd480e
- Chromosome Location
- Chr. 16: 44347395 - 44388492 on Build GRCm38
- UniGene ID
- Mm.24035
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144550.4 |
NP_653133.3
|
18 |
3218 |
|
GenBank mRNA |
AK048789.1 |
- |
18 |
3162 |
|
View Full Product Details