Gene Transcripts

Gene Symbol
Btbd3
Entrez Gene ID
228662
Gene Name
BTB (POZ) domain containing 3
Gene Aliases
mKIAA0952
Chromosome Location
Chr. 2: 138256584 - 138287424 on Build GRCm38
UniGene ID
Mm.153405
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025431.1 NP_001020602.1 2, 3 459
NM_145534.2 NP_663509.2 1, 2 761
GenBank mRNA AB093275.1 - 1, 2 761
AK052530.1 - 2, 3 420
AK161924.1 - 2, 3 416
BC018174.1 - 1, 2 738 4011475
BC062968.1 - 1, 2 457 6408511

View Full Product Details