Gene Transcripts

Gene Symbol
Trim31
Entrez Gene ID
224762
Gene Name
tripartite motif-containing 31
Gene Aliases
BC026666, HCG1, HCGI, RNF
Chromosome Location
Chr. 17: 36898130 - 36910217 on Build GRCm38
UniGene ID
Mm.213417
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146077.2 NP_666189.1 1 436
GenBank mRNA AK033650.1 - 1 449
BC026666.1 - 1 436 4990292

View Full Product Details