Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005780.2 | NP_005771.1 | 4 | 1477 | |
XM_011534861.1 | XP_011533163.1 | 4 | 1280 | ||
GenBank mRNA | AF098807.1 | - | 4 | 1316 | |
AK129819.1 | - | 4 | 973 | ||
AK223369.1 | - | 4 | 973 | ||
AL137388.1 | - | 1 | 144 |