Gene Transcripts

Gene Symbol
Uba6
Entrez Gene ID
231380
Gene Name
ubiquitin-like modifier activating enzyme 6
Gene Aliases
4930542H01, 5730469D23Rik, AU021846, AW124799, E1-L2, Ube1l2
Chromosome Location
Chr. 5: 86110720 - 86172743 on Build GRCm38
UniGene ID
Mm.34012
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172712.2 NP_766300.1 33 3320
GenBank mRNA AK029769.1 - 33 3352
AK049603.1 - 33 3358
BC063048.1 - 33 3320 6825308

View Full Product Details