Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001048192.2 | NP_001041657.1 | 8 | 1153 | |
NM_001294262.1 | NP_001281191.1 | 7 | 1181 | ||
NM_174849.3 | NP_777274.1 | 7 | 1070 | ||
GenBank mRNA | AK029676.1 | - | 7 | 1071 | |
AK035680.1 | - | 5 | 638 | ||
AK044195.1 | - | 6 | 2475 | ||
AK083656.1 | - | 7 | 1020 | ||
BC057349.1 | - | 7 | 1071 | 6813901 | |
DQ867034.1 | - | 7 | 1066 | ||
DQ867035.1 | - | 7 | 1066 | ||
DQ867036.1 | - | 8 | 1153 | ||
DQ867037.1 | - | 7 | 1066 | ||
DQ867038.1 | - | 7 | 1066 |