Gene Transcripts

Gene Symbol
Zcchc14
Entrez Gene ID
142682
Gene Name
zinc finger, CCHC domain containing 14
Gene Aliases
AA792890, BDG29
Chromosome Location
Chr. 8: 121598697 - 121652761 on Build GRCm38
UniGene ID
Mm.103202
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080855.2 NP_543131.1 5 571
GenBank mRNA AB030244.1 - 5 582
BC132449.1 - 5 569 40130726
BC137853.1 - 5 569 8860872
CB518791.1 - 3 217
CJ102007.1 - 5 414

View Full Product Details