Gene Transcripts

Gene Symbol
Akr1c14
Entrez Gene ID
105387
Gene Name
aldo-keto reductase family 1, member C14
Gene Aliases
9030611N15Rik, AW557061
Chromosome Location
Chr. 13: 4048999 - 4090560 on Build GRCm38
UniGene ID
Mm.26838
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134072.1 NP_598833.1 9 1183
GenBank mRNA AK033537.1 - 9 1201
AK136528.1 - 9 1200
BC013482.1 - 9 1183 4240772
BC026628.1 - 9 1182 4983237

View Full Product Details