Gene Transcripts

Gene Symbol
Bspry
Entrez Gene ID
192120
Gene Name
B-box and SPRY domain containing
Gene Aliases
Chromosome Location
Chr. 4: 62479995 - 62497300 on Build GRCm38
UniGene ID
Mm.29808
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138653.1 NP_619594.1 7 1699
GenBank mRNA AJ276690.1 - 7 1699
AK035588.1 - 6 2115
AK160930.1 - 6 2115
BC003992.1 - 4 1652

View Full Product Details