Gene Transcripts

Gene Symbol
Fam64a
Entrez Gene ID
109212
Gene Name
family with sequence similarity 64, member A
Gene Aliases
2610008F03Rik, 6720460F02Rik, AI115087, CATS
Chromosome Location
Chr. 11: 72042439 - 72047371 on Build GRCm38
UniGene ID
Mm.33118
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144526.3 NP_653109.2 6 863
GenBank mRNA AK011345.1 - 6 861
AK049710.1 - 6 864
AK078429.1 - 6 864

View Full Product Details