Gene Transcripts
- Gene Symbol
- UBE4B
- Entrez Gene ID
- 10277
- Gene Name
- ubiquitination factor E4B
- Gene Aliases
- E4, HDNB1, UBOX3, UFD2, UFD2A
- Chromosome Location
- Chr. 1: 10032983 - 10181239 on Build GRCh38
- UniGene ID
- Hs.593974
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001105562.2 |
NP_001099032.1
|
14, 15 |
2606 |
|
NM_006048.4 |
NP_006039.2
|
13, 14 |
2219 |
|
XM_005263422.1 |
XP_005263479.1
|
15, 16 |
2583 |
|
XM_011540488.2 |
XP_011538790.1
|
14, 15 |
1882 |
|
XM_011540489.2 |
XP_011538791.1
|
15, 16 |
2583 |
|
XM_011540490.1 |
XP_011538792.1
|
15, 16 |
2583 |
|
XM_017000035.1 |
XP_016855524.1
|
12, 13 |
1669 |
|
XM_017000036.1 |
XP_016855525.1
|
14, 15 |
1883 |
|
GenBank mRNA |
AB014584.3 |
- |
13, 14 |
2110 |
|
AB028839.1 |
- |
13, 14 |
2227 |
|
AF043117.1 |
- |
14, 15 |
2003 |
|
AF331520.1 |
- |
13, 14 |
1578 |
|
AK023654.1 |
- |
3, 4 |
1333 |
|
AK292444.1 |
- |
13, 14 |
1701 |
|
AK304484.1 |
- |
11, 12 |
1206 |
|
BC093696.1 |
- |
13, 14 |
1609 |
7939541 |
BC143458.1 |
- |
13, 14 |
1609 |
|
BC143460.1 |
- |
12, 13 |
1494 |
|
JF289274.1 |
- |
15, 16 |
2071 |
|
View Full Product Details