Gene Transcripts

Gene Symbol
Ldhal6b
Entrez Gene ID
106557
Gene Name
lactate dehydrogenase A-like 6B
Gene Aliases
4933402O15Rik, AI326310, LDHL, Ldhal
Chromosome Location
Chr. 17: 5417323 - 5418767 on Build GRCm38
UniGene ID
Mm.432764
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175349.2 NP_780558.1 1 736
GenBank mRNA AK077089.1 - 1 682
AY280363.1 - 1 736
BC061193.1 - 1 681 6704800

View Full Product Details