Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161844.1 | NP_001155316.1 | 6 | 1020 | |
NM_023224.5 | NP_075713.3 | 6 | 1020 | ||
GenBank mRNA | AF319956.1 | - | 6 | 993 | |
AK010228.1 | - | 6 | 1021 | ||
BC046337.1 | - | 6 | 996 | 6395806 | |
BC111037.1 | - | 6 | 1004 | 5353413 | |
BE198430.1 | - | 4 | 363 |