Gene Transcripts
- Gene Symbol
- Pkd1l2
- Entrez Gene ID
- 76645
- Gene Name
- polycystic kidney disease 1 like 2
- Gene Aliases
- 1700126L06Rik
- Chromosome Location
- Chr. 8: 116995218 - 117082488 on Build GRCm38
- UniGene ID
- Mm.159753
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_029686.4 |
NP_083962.4
|
22 |
3552 |
|
GenBank mRNA |
AY164484.1 |
- |
22 |
3555 |
|
View Full Product Details