Gene Transcripts

Gene Symbol
Smarcd1
Entrez Gene ID
83797
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1
Gene Aliases
AA407987, Baf60a, D15Kz1
Chromosome Location
Chr. 15: 99702287 - 99713995 on Build GRCm38
UniGene ID
Mm.273756
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031842.2 NP_114030.2 13 1870
GenBank mRNA AK075611.1 - 9 1206
AK187748.1 - 1 47
BC026783.1 - 13 1782 4481097
BC059921.1 - 13 1840
BC079839.1 - 13 1816 6810462
M25773.1 - 13 1820
U66620.1 - 13 1822

View Full Product Details