Gene Transcripts

Gene Symbol
Sval2
Entrez Gene ID
84543
Gene Name
seminal vesicle antigen-like 2
Gene Aliases
SLP-M
Chromosome Location
Chr. 6: 41860339 - 41864322 on Build GRCm38
UniGene ID
Mm.138766
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032542.1 NP_115931.1 3 257
GenBank mRNA AB050614.1 - 3 257
BC119343.1 - 3 357 8734092
BC119345.1 - 3 357 8734094

View Full Product Details