Gene Transcripts

Gene Symbol
Hscb
Entrez Gene ID
100900
Gene Name
HscB iron-sulfur cluster co-chaperone homolog (E. coli)
Gene Aliases
AI325508, AW049829, Hsc20
Chromosome Location
Chr. 5: 110829070 - 110839802 on Build GRCm38
UniGene ID
Mm.200231
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153571.2 NP_705799.2 2, 3 374
GenBank mRNA AK131961.1 - 2, 3 376
BC027641.1 - 2, 3 338 1244839

View Full Product Details