Gene Transcripts

Gene Symbol
Ccdc12
Entrez Gene ID
72654
Gene Name
coiled-coil domain containing 12
Gene Aliases
2700094L05Rik, C76605
Chromosome Location
Chr. 9: 110656503 - 110711593 on Build GRCm38
UniGene ID
Mm.249115
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028312.3 NP_082588.2 7 680
GenBank mRNA AK012610.1 - 7 681
BC026668.1 - 7 630 5006213
CA450868.1 - 5 414

View Full Product Details