Gene Transcripts

Gene Symbol
Mettl13
Entrez Gene ID
71449
Gene Name
methyltransferase like 13
Gene Aliases
5630401D24Rik
Chromosome Location
Chr. 1: 162533671 - 162548345 on Build GRCm38
UniGene ID
Mm.270569
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144877.1 NP_659126.1 1 179
GenBank mRNA AK017470.1 - 1 364
AK048636.1 - 1 362
AK050492.1 - 1 372
AK084750.1 - 1 357
AK146751.1 - 377
AK162843.1 - 1 364
AK168959.1 - 1 386
AK169319.1 - 378
AK173044.1 - 332
BC014872.1 - 1 179 3602469

View Full Product Details