Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006453.2 | NP_006444.2 | 4 | 308 | |
GenBank mRNA | AB209173.1 | - | 4 | 280 | |
BC010231.1 | - | 4 | 308 | 3455740 | |
BC014007.2 | - | 4 | 280 | 4040312 | |
BC035409.1 | - | 4 | 264 | 5165833 | |
U02609.1 | - | 4 | 258 |