Gene Transcripts

Gene Symbol
Zcrb1
Entrez Gene ID
67197
Gene Name
zinc finger CCHC-type and RNA binding motif 1
Gene Aliases
2700088M22Rik, Madp-1
Chromosome Location
Chr. 15: 93386106 - 93398329 on Build GRCm38
UniGene ID
Mm.293181
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026025.2 NP_080301.1 5 389
GenBank mRNA AB095989.1 - 5 422
AK012589.1 - 6 472
AK012852.1 - 5 390
AK170136.1 - 5 401
BC058368.1 - 5 423 6819507

View Full Product Details