Gene Transcripts

Gene Symbol
Pbld2
Entrez Gene ID
67307
Gene Name
phenazine biosynthesis-like protein domain containing 2
Gene Aliases
3110049J23Rik, Mawbp
Chromosome Location
Chr. 10: 63024049 - 63058813 on Build GRCm38
UniGene ID
Mm.368563
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026085.2 NP_080361.1 10 1878
GenBank mRNA AK014198.1 - 10 1879
AK018692.1 - 9 1634
BC061073.1 - 9 1633 30381600

View Full Product Details