Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027399.3 | NP_081675.2 | 5 | 1123 | |
GenBank mRNA | AF297098.1 | - | 4 | 1053 | |
AK010437.1 | - | 5 | 1118 | ||
AK144312.1 | - | 5 | 1078 | ||
AY029584.1 | - | 5 | 1082 | ||
BC061023.1 | - | 5 | 1108 | 30304473 |