Gene Transcripts

Gene Symbol
Tmem97
Entrez Gene ID
69071
Gene Name
transmembrane protein 97
Gene Aliases
1810014L12Rik, AI115531, AL022956, D11Bhm182e
Chromosome Location
Chr. 11: 78541817 - 78550735 on Build GRCm38
UniGene ID
Mm.29431
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133706.2 NP_598467.1 3 403
GenBank mRNA AK007497.1 - 2 132
AK155107.1 - 3 415
AK171954.1 - 3 382
BC018156.1 - 3 403 4192256
BC093511.1 - 3 380 5124076

View Full Product Details