Gene Transcripts

Gene Symbol
CD226
Entrez Gene ID
10666
Gene Name
CD226 molecule
Gene Aliases
DNAM-1, DNAM1, PTA1, TLiSA1
Chromosome Location
Chr. 18: 69860228 - 69962086 on Build GRCh38
UniGene ID
Hs.660130
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303618.1 NP_001290547.1 6 2638
NM_001303619.1 NP_001290548.1 5 2231
NM_006566.3 NP_006557.2 7 2640
XM_005266642.3 XP_005266699.1 7 2739
XM_005266643.3 XP_005266700.1 5 1833
XM_006722374.3 XP_006722437.1 5 2304
XM_017025524.1 XP_016881013.1 7 9383
XM_017025525.1 XP_016881014.1 5 2115
XM_017025526.1 XP_016881015.1 6 2009
XM_017025527.1 XP_016881016.1 5 2673
GenBank mRNA GU935335.1 - 7 2387
GU935336.1 - 5 1990
U56102.1 - 7 2399

View Full Product Details