Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023048.5 | NP_075535.1 | 2, 3 | 225 | |
GenBank mRNA | AF155355.1 | - | 1, 2 | 195 | |
AK018373.1 | - | 1, 2 | 378 | ||
AK132068.1 | - | 1, 2 | 387 | ||
AK133894.1 | - | 1, 2 | 96 | ||
BC046819.1 | - | 2, 3 | 225 | 5703017 | |
BC054745.1 | - | 1, 2 | 333 | 6836490 | |
BC056440.1 | - | 2, 3 | 225 | 5703017 |