Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025321.3 | NP_079597.2 | 3 | 170 | |
GenBank mRNA | AK002459.1 | - | 3 | 172 | |
AK012818.1 | - | 3 | 171 | ||
AK032458.1 | - | 172 | |||
AK135503.1 | - | 3 | 172 | ||
BC005779.1 | - | 3 | 157 | 2810852 | |
BC083091.1 | - | 3 | 163 | 30445339 | |
BY711932.1 | - | 3 | 171 |