Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042513.1 | NP_001035978.1 | 11 | 1263 | |
NM_001042514.1 | NP_001035979.1 | 10 | 1351 | ||
NM_001042523.1 | NP_001035988.1 | 11 | 1480 | ||
NM_015762.2 | NP_056577.2 | 10 | 1156 | ||
GenBank mRNA | AB027565.1 | - | 10 | 1183 | |
AF333036.1 | - | 11 | 1480 | ||
AK146125.1 | - | 10 | 1157 | ||
AK149625.1 | - | 11 | 1264 | ||
AK151927.1 | - | 11 | 1263 | ||
AK168356.1 | - | 11 | 1264 | ||
AK181192.1 | - | 4 | 366 | ||
AK183038.1 | - | 4 | 367 | ||
AK201250.1 | - | 1 | 87 | ||
BC020274.1 | - | 10 | 1118 | ||
BC037643.1 | - | 10 | 1436 | 5059831 |